Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002199.3 | NP_002190.2 | 9 | 1362 | ||
GenBank mRNA | BC015803.2 | - | 8 | 1169 | 3920890 | |
BX648934.1 | - | 1 | 5935 | |||
X15949.1 | - | 9 | 1260 |