Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080622.3 | NP_542189.1 | 1 | 1227 | ||
GenBank mRNA | BC031949.1 | - | 1 | 942 |