Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CBLB SIRNA Assays ›
Gene Symbol:
CBLB
Gene Name:
Cbl proto-oncogene B
Gene Aliases:
Cbl-b, Nbla00127, RNF56
Chromosome Location:
Chr.3: 105655461 - 105869552 on Build GRCh38
UniGene ID:
Hs.430589
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43983
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CBLB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321786.1
NP_001308715.1
17
2721
NM_001321788.1
NP_001308717.1
17
2640
NM_001321789.1
NP_001308718.1
16
2512
NM_001321790.1
NP_001308719.1
16
2462
NM_001321791.1
NP_001308720.1
16
2508
NM_001321793.1
NP_001308722.1
16
2684
NM_001321794.1
NP_001308723.1
16
2493
NM_001321795.1
NP_001308724.1
16
2669
NM_001321796.1
NP_001308725.1
16
2493
NM_001321797.1
NP_001308726.1
15
2361
NM_001321798.1
NP_001308727.1
15
2537
NM_001321799.1
NP_001308728.1
15
2537
NM_001321806.1
NP_001308735.1
16
2483
NM_001321807.1
NP_001308736.1
18
2960
NM_001321808.1
NP_001308737.1
17
2652
NM_001321811.1
NP_001308740.1
15
2512
NM_001321813.1
NP_001308742.1
17
2550
NM_001321816.1
NP_001308745.1
14
2380
NM_001321820.1
NP_001308749.1
18
2816
NM_001321822.1
NP_001308751.1
18
2866
NM_170662.4
NP_733762.2
17
2816
NR_135806.1
18
2948
NR_135807.1
18
2864
NR_135808.1
17
2816
NR_135809.1
17
2798
NR_135810.1
17
2594
NR_135811.1
17
2640
NR_135812.1
16
2404
XM_011513257.1
XP_011511559.1
17
2636
XM_011513259.2
XP_011511561.2
17
2761
XM_017007395.1
XP_016862884.1
17
2588
XM_017007396.1
XP_016862885.1
16
2627
XM_017007397.1
XP_016862886.1
16
2613
XM_017007398.1
XP_016862887.1
16
2439
XM_017007399.1
XP_016862888.1
16
2664
XM_017007400.1
XP_016862889.1
15
2481
GenBank mRNA
AB075490.1
-
6
1099
AK094184.1
-
8
1042
AK123147.1
-
2
473
AK292792.1
-
17
2757
AK302533.1
-
16
2462
AK316276.1
-
16
2442
BC032851.2
-
17
2757
5269036
CR749655.1
-
5
511
DQ349203.1
-
16
2494
DQ349204.1
-
16
2494
U26710.1
-
17
2816
Back To Top
More Information
Panther Classification:
Molecular Function -
damaged DNA-binding protein
ligase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
NLS-bearing protein import into nucleus
signal transduction
cell surface receptor signaling pathway
response to gravity
response to hormone
regulation of signaling
phosphotyrosine binding
ubiquitin-protein transferase activity
signal transducer activity
calcium ion binding
protein binding
zinc ion binding
ligase activity
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0