Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NINJ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294345.1 | NP_001281274.1 | 3 | 365 | ||
NM_001294346.1 | NP_001281275.1 | 3 | 472 | |||
NM_016533.5 | NP_057617.2 | 3 | 711 | |||
GenBank mRNA | AF205633.1 | - | 3 | 386 | ||
BC057766.1 | - | 3 | 379 | |||
BI597990.1 | - | 3 | 564 | |||
BM128205.1 | - | 3 | 333 | |||
CD695373.1 | - | 3 | 459 |