Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173833.5 | NP_776194.2 | 7 | 1742 | ||
GenBank mRNA | AK172746.1 | - | 7 | 1572 | ||
AK301006.1 | - | 5 | 383 | |||
AY358150.1 | - | 6 | 890 | |||
BC033153.1 | - | 7 | 1514 | 4138639 |