Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164522.2 | NP_001157994.1 | 3 | 343 | ||
NM_001304839.1 | NP_001291768.1 | 3 | 343 | |||
NM_177403.5 | NP_796377.3 | 2 | 417 | |||
XM_006711288.3 | XP_006711351.1 | 3 | 340 | |||
GenBank mRNA | AK122657.1 | - | 3 | 338 | ||
AK127140.1 | - | 2 | 401 | |||
AY094596.1 | - | 3 | 290 | |||
BC017092.1 | - | 3 | 285 | 3851998 | ||
BI757251.1 | - | 2 | 414 | |||
DA966224.1 | - | 3 | 343 |