Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177424.2 | NP_803173.1 | 4 | 514 | ||
GenBank mRNA | AF123769.1 | - | 4 | 461 | ||
AK297585.1 | - | 4 | 442 | |||
AK299982.1 | - | 4 | 346 | |||
AK309990.1 | - | 3 | 217 | |||
AK315880.1 | - | 2 | 145 | |||
AL035306.1 | - | 4 | 457 | |||
BC046999.1 | - | 4 | 442 | 3851266 | ||
CR407620.1 | - | 4 | 368 |