Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLT8D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010983.2 | NP_001010983.1 | 4 | 993 | ||
NM_001278280.1 | NP_001265209.1 | 4 | 989 | |||
NM_001278281.1 | NP_001265210.1 | 4 | 737 | |||
NM_018446.3 | NP_060916.1 | 3 | 764 | |||
NM_152932.2 | NP_690909.1 | 3 | 322 | |||
XM_017006857.1 | XP_016862346.1 | 5 | 1152 | |||
XM_017006858.1 | XP_016862347.1 | 4 | 923 | |||
XM_017006859.1 | XP_016862348.1 | 3 | 472 | |||
GenBank mRNA | AF157318.1 | - | 3 | 209 | ||
AF175227.1 | - | 4 | 476 | |||
AF176777.1 | - | 4 | 475 | |||
AJ271710.1 | - | 2 | 91 | |||
AK027517.1 | - | 3 | 230 | |||
AK075031.1 | - | 2 | 286 | |||
AY358579.1 | - | 3 | 243 | |||
BC119670.1 | - | 3 | 208 | 40116196 | ||
BC119671.1 | - | 3 | 208 | 40116197 | ||
BF725274.1 | - | 4 | 410 | |||
BG700867.1 | - | 4 | 679 | |||
CR749821.1 | - | 4 | 446 |