Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2AFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012412.4 | NP_036544.1 | 3 | 267 | ||
NM_138635.3 | NP_619541.1 | 3 | 267 | |||
NM_201436.2 | NP_958844.1 | 2 | 189 | |||
NM_201516.2 | NP_958924.1 | 3 | 267 | |||
GenBank mRNA | AB209001.1 | - | 3 | 178 | ||
AF081192.1 | - | 3 | 132 | |||
AK023973.1 | - | 3 | 218 | |||
AK129989.1 | - | 3 | 187 | |||
AL520811.3 | - | 3 | 168 | |||
BC000098.2 | - | 3 | 194 | 3505930 | ||
BC004274.1 | - | 2 | 119 | 3615320 | ||
BC014885.2 | - | 3 | 163 | 3907411 | ||
BC070169.1 | - | 3 | 194 | 6458566 | ||
BF242439.1 | - | 3 | 281 | |||
BG754026.1 | - | 3 | 242 | |||
BG773256.1 | - | 3 | 270 | |||
BI257891.1 | - | 3 | 225 | |||
BM452844.1 | - | 3 | 148 | |||
BM453663.1 | - | 3 | 137 | |||
CB148943.1 | - | 3 | 221 | |||
DB460745.1 | - | 3 | 267 |