Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD54B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205263.1 | NP_001192192.1 | 2 | 484 | ||
NM_012415.3 | NP_036547.1 | 4 | 461 | |||
GenBank mRNA | AF112481.1 | - | 4 | 410 | ||
AK290437.1 | - | 4 | 431 | |||
AK307516.1 | - | 2 | 157 | |||
BC001965.2 | - | 4 | 403 | 3452509 | ||
BC020668.1 | - | 5 | 474 | |||
BP872374.1 | - | 4 | 461 | |||
DA553267.1 | - | 2 | 484 |