Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002709.2 | NP_002700.1 | 8 | 1363 | ||
NM_206876.1 | NP_996759.1 | 9 | 1158 | |||
GenBank mRNA | AF092905.1 | - | 10 | 1136 | ||
AK316532.1 | - | 8 | 984 | |||
BC002697.2 | - | 8 | 1079 | 3609386 | ||
BC012045.1 | - | 8 | 1079 | 3631338 | ||
BX647295.1 | - | 4 | 554 | |||
BX647970.1 | - | 9 | 1197 | |||
EU668339.1 | - | 8 | 1079 | |||
X80910.1 | - | 8 | 1256 |