Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033053.2 | NP_001028225.1 | 1 | 638 | ||
NM_014922.4 | NP_055737.1 | 1 | 638 | |||
NM_033004.3 | NP_127497.1 | 1 | 638 | |||
NM_033006.3 | NP_127499.1 | 1 | 638 | |||
NM_033007.3 | NP_127500.1 | 1 | 638 | |||
GenBank mRNA | AB023143.1 | - | 1 | 605 | ||
AF229059.1 | - | 1 | 83 | |||
AF229060.1 | - | 1 | 83 | |||
AF229061.1 | - | 1 | 83 | |||
AF229062.1 | - | 1 | 83 | |||
AF298548.1 | - | 2 | 242 | |||
AF310105.1 | - | 1 | 601 | |||
AK310936.1 | - | 2 | 449 |