Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025138.4 | NP_079414.3 | 11 | 1712 | ||
XM_005266545.3 | XP_005266602.1 | 10 | 1685 | |||
XM_011535239.2 | XP_011533541.1 | 10 | 1645 | |||
GenBank mRNA | AB107354.1 | - | 11 | 1712 | ||
AK022723.1 | - | 5 | 388 | |||
AK074360.1 | - | 11 | 1684 | |||
BC053566.1 | - | 5 | 391 | |||
BC064348.1 | - | 11 | 967 | 4939198 | ||
BX647471.1 | - | 11 | 1134 |