Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130773.3 | NP_570129.1 | 3 | 654 | ||
XM_006712258.1 | XP_006712321.1 | 3 | 654 | |||
XM_017003316.1 | XP_016858805.1 | 3 | 654 | |||
GenBank mRNA | AB077881.1 | - | 3 | 290 | ||
AK056528.1 | - | 3 | 654 |