Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014507.3 | NP_055322.1 | 2 | 489 | ||
NM_173467.4 | NP_775738.3 | 2 | 489 | |||
NR_046423.1 | 2 | 523 | ||||
XM_011530156.2 | XP_011528458.1 | 2 | 103 | |||
GenBank mRNA | AA402283.1 | - | 2 | 53 | ||
AK314059.1 | - | 2 | 471 | |||
AL359401.1 | - | 2 | 456 | |||
AL359403.1 | - | 2 | 456 | |||
BC030985.1 | - | 2 | 441 | |||
BC042195.1 | - | 2 | 482 | 6052380 | ||
BI464466.1 | - | 2 | 493 |