Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PILRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178238.3 | NP_839956.1 | 3 | 828 | ||
GenBank mRNA | AF161081.1 | - | 3 | 816 | ||
AJ400845.1 | - | 3 | 836 | |||
BC050547.1 | - | 6 | 875 | 5755993 |
Gene Symbol: STAG3L5P-PVRIG2P-PILRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036569.1 | 17 | 3073 | |||
NR_036570.1 | 16 | 2397 | ||||
GenBank mRNA | AF217981.1 | - | 8 | 1802 | ||
AJ400846.1 | - | 6 | 913 | |||
AK056412.1 | - | 17 | 3024 | |||
AK298676.1 | - | 4 | 866 | |||
AL834317.1 | - | 16 | 2219 | |||
AL834336.1 | - | 4 | 742 |