Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277126.1 | NP_001264055.1 | 9 | 3195 | ||
NM_001277129.1 | NP_001264058.1 | 8 | 3021 | |||
NM_144687.3 | NP_653288.1 | 9 | 3192 | |||
XM_011527479.1 | XP_011525781.1 | 8 | 3024 | |||
XM_011527480.1 | XP_011525782.1 | 8 | 3024 | |||
XM_011527482.1 | XP_011525784.1 | 7 | 2853 | |||
XM_017027460.1 | XP_016882949.1 | 9 | 3192 | |||
XM_017027462.1 | XP_016882951.1 | 8 | 3021 | |||
XM_017027463.1 | XP_016882952.1 | 10 | 2968 | |||
XM_017027464.1 | XP_016882953.1 | 9 | 2766 | |||
XM_017027465.1 | XP_016882954.1 | 10 | 2976 | |||
XM_017027466.1 | XP_016882955.1 | 10 | 2973 | |||
XM_017027467.1 | XP_016882956.1 | 9 | 2768 | |||
GenBank mRNA | AF231021.1 | - | 6 | 1949 | ||
AK095460.1 | - | 8 | 2936 | |||
AK290772.1 | - | 9 | 2974 | |||
AY095146.1 | - | 9 | 3139 | |||
AY116204.1 | - | 9 | 3186 | |||
AY116206.1 | - | 7 | 2850 | |||
AY154467.1 | - | 9 | 3139 | |||
BC028069.1 | - | 9 | 3082 | 5212737 |