Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYKPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278346.1 | NP_001265275.1 | 11 | 2320 | ||
NM_153373.3 | NP_699204.1 | 11 | 1497 | |||
NR_103508.1 | 8 | 1083 | ||||
XM_006714925.2 | XP_006714988.1 | 10 | 1333 | |||
XM_011534669.2 | XP_011532971.1 | 11 | 1489 | |||
XM_011534670.2 | XP_011532972.1 | 10 | 1822 | |||
XM_011534671.2 | XP_011532973.1 | 11 | 1477 | |||
XM_011534672.2 | XP_011532974.1 | 11 | 1275 | |||
XM_011534674.2 | XP_011532976.1 | 8 | 1144 | |||
XM_011534676.2 | XP_011532978.1 | 7 | 1429 | |||
XM_017009998.1 | XP_016865487.1 | 10 | 1791 | |||
XM_017009999.1 | XP_016865488.1 | 7 | 3234 | |||
GenBank mRNA | AF318375.1 | - | 4 | 2434 | ||
AK023470.1 | - | 8 | 1449 | |||
AK093737.1 | - | 6 | 1171 | |||
AK292061.1 | - | 11 | 1459 | |||
BC008009.1 | - | 8 | 1051 | |||
BC037567.1 | - | 11 | 1459 | 5220434 | ||
BC067745.1 | - | 10 | 1168 | |||
BC110335.1 | - | 11 | 1469 | 5753177 |