Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193451.1 | NP_001180380.1 | 11 | 1783 | ||
NM_175861.3 | NP_787057.2 | 11 | 1874 | |||
XM_005253499.4 | XP_005253556.1 | 13 | 2035 | |||
XM_017020003.1 | XP_016875492.1 | 12 | 1868 | |||
XM_017020004.1 | XP_016875493.1 | 12 | 1828 | |||
XM_017020005.1 | XP_016875494.1 | 10 | 1641 | |||
XM_017020006.1 | XP_016875495.1 | 13 | 2039 | |||
XM_017020007.1 | XP_016875496.1 | 9 | 1061 | |||
GenBank mRNA | AK055962.1 | - | 9 | 1035 | ||
AK123619.1 | - | 7 | 1465 | |||
AK127297.1 | - | 13 | 1845 | |||
AL832328.1 | - | 2 | 199 | |||
BC042083.1 | - | 11 | 1874 | 5749757 | ||
GQ865601.1 | - | 11 | 1783 |