Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003063.2 | NP_003054.1 | 2 | 262 | ||
GenBank mRNA | BC005261.1 | - | 2 | 257 | 3934241 | |
BC094685.1 | - | 2 | 244 | 6203008 | ||
BC104150.2 | - | 2 | 214 | 40029313 | ||
BC104185.2 | - | 2 | 209 | 40030832 | ||
BC113930.1 | - | 2 | 213 | 40030834 | ||
BC113987.1 | - | 2 | 191 | 40030833 | ||
BP233121.1 | - | 2 | 262 | |||
U96094.1 | - | 2 | 256 |