Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032779.3 | NP_116168.3 | 9 | 2390 | ||
GenBank mRNA | AK027303.1 | - | 3 | 588 | ||
AK075543.1 | - | 9 | 2156 | |||
BC117269.1 | - | 9 | 2108 | 40125820 | ||
BC143398.1 | - | 9 | 2108 | 9051905 | ||
BC143399.1 | - | 8 | 2221 |