Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039661.1 | NP_001034750.1 | 5 | 680 | ||
NM_001318776.1 | NP_001305705.1 | 4 | 527 | |||
NM_001318777.1 | NP_001305706.1 | 4 | 527 | |||
NM_148910.2 | NP_683708.1 | 5 | 696 | |||
XM_005271399.3 | XP_005271456.2 | 3 | 425 | |||
XM_011542576.2 | XP_011540878.1 | 3 | 425 | |||
XM_017017161.1 | XP_016872650.1 | 2 | 2048 | |||
XM_017017162.1 | XP_016872651.1 | 5 | 717 | |||
GenBank mRNA | AB446477.1 | - | 2 | 244 | ||
AF378129.1 | - | 5 | 677 | |||
AF406652.1 | - | 2 | 244 | |||
AF410783.1 | - | 5 | 673 | |||
AK124298.1 | - | 4 | 527 | |||
AK313147.1 | - | 4 | 520 | |||
AY576785.1 | - | 2 | 283 | |||
AY576786.1 | - | 2 | 283 | |||
AY576787.1 | - | 2 | 283 | |||
BC032474.1 | - | 5 | 649 | 5216171 |