Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185009.2 | NP_001171938.1 | 7 | 1108 | ||
NM_002938.4 | NP_002929.1 | 6 | 586 | |||
GenBank mRNA | AB000468.1 | - | 6 | 535 | ||
AK128038.1 | - | 6 | 1749 | |||
AK312534.1 | - | 6 | 515 | |||
BC031935.1 | - | 7 | 1108 | 4824114 | ||
U95140.1 | - | 6 | 508 |