Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000202.7 | NP_000193.1 | 2, 3 | 450 | ||
NM_006123.4 | NP_006114.1 | 2, 3 | 450 | |||
NR_104128.1 | 2, 3 | 450 | ||||
GenBank mRNA | AF050145.1 | - | 2, 3 | 251 | ||
BC006170.1 | - | 2, 3 | 308 | 4025980 | ||
DA325538.1 | - | 2, 3 | 450 | |||
DB500211.1 | - | 2, 3 | 435 | |||
L40586.1 | - | 2, 3 | 360 | |||
M58342.1 | - | 2, 3 | 357 |