Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000980.3 | NP_000971.1 | 2 | 210 | ||
GenBank mRNA | AK222647.1 | - | 2 | 166 | ||
AK297323.1 | - | 2 | 167 | |||
AK297358.1 | - | 2 | 165 | |||
AK300809.1 | - | 1 | 162 | |||
AK311774.1 | - | 2 | 166 | |||
BC007512.2 | - | 1 | 150 | 2961519 | ||
BC066319.1 | - | 1 | 170 | 5286436 | ||
BC071920.1 | - | 1 | 150 | 6293838 | ||
BI753938.1 | - | 153 | ||||
HY050156.1 | - | 2 | 211 | |||
L05093.1 | - | 2 | 150 | |||
X80822.1 | - | 1 | 157 |