Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SECTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003004.2 | NP_002995.1 | 2, 3 | 486 | ||
XM_005256392.3 | XP_005256449.1 | 2, 3 | 270 | |||
XM_011523588.2 | XP_011521890.1 | 2, 3 | 517 | |||
GenBank mRNA | AK223068.1 | - | 2, 3 | 179 | ||
AK223104.1 | - | 2, 3 | 210 | |||
AK290708.1 | - | 2, 3 | 439 | |||
AK312980.1 | - | 2, 3 | 206 | |||
BC017716.1 | - | 2, 3 | 180 | 4413387 | ||
DA589733.1 | - | 2 | 486 | |||
U77643.1 | - | 2, 3 | 206 |