Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317165.1 | NP_001304094.1 | 3 | 204 | ||
NM_001317166.1 | NP_001304095.1 | 3 | 204 | |||
NM_001317167.1 | NP_001304096.1 | 3 | 122 | |||
NM_001317168.1 | NP_001304097.1 | 3 | 834 | |||
NM_001317169.1 | NP_001304098.1 | 4 | 247 | |||
NM_001317171.1 | NP_001304100.1 | 2 | 181 | |||
NM_003096.3 | NP_003087.1 | 3 | 204 | |||
XM_017004774.1 | XP_016860263.1 | 3 | 163 | |||
GenBank mRNA | AV682126.1 | - | 3 | 204 | ||
BC000070.2 | - | 1 | 137 | 3503973 | ||
BC022432.1 | - | 3 | 170 | 4276853 | ||
BC035103.1 | - | 3 | 815 | 5262406 | ||
BC066302.1 | - | 1 | 127 | 30336948 | ||
BC070166.1 | - | 1 | 120 | 5809503 | ||
BC071880.1 | - | 1 | 94 | 3640165 | ||
BC106055.1 | - | 3 | 187 | 4367206 | ||
BP424287.1 | - | 3 | 144 | |||
BU540400.1 | - | 3 | 173 | |||
CD109295.1 | - | 2 | 150 | |||
CR456918.1 | - | 3 | 82 | |||
DW428820.1 | - | 3 | 209 | |||
HY168189.1 | - | 4 | 248 | |||
X85373.1 | - | 3 | 165 |
Gene Symbol: SNRPGP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BF085802.1 | - | 1 | 198 | ||
BF367571.1 | - | 1 | 173 |