Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000023.3 | NP_000014.1 | 10 | 1381 | ||
NM_001135697.2 | NP_001129169.1 | 8 | 1009 | |||
NR_135553.1 | 9 | 1172 | ||||
XM_011525120.2 | XP_011523422.2 | 10 | 1773 | |||
XM_011525121.2 | XP_011523423.2 | 10 | 1625 | |||
XM_011525122.2 | XP_011523424.2 | 9 | 1564 | |||
XM_011525123.2 | XP_011523425.2 | 8 | 1401 | |||
XM_011525124.2 | XP_011523426.1 | 9 | 1128 | |||
GenBank mRNA | BC025702.1 | - | 10 | 1353 | 5222483 | |
F25770.1 | - | 2 | 323 | |||
L34355.1 | - | 10 | 1362 | |||
L35853.1 | - | 8 | 990 | |||
U08895.1 | - | 10 | 1336 |