Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284367.1 | NP_001271296.1 | 2 | 239 | ||
NM_001284368.1 | NP_001271297.1 | 2 | 148 | |||
NM_005004.3 | NP_004995.1 | 2 | 239 | |||
GenBank mRNA | AF044958.1 | - | 2 | 189 | ||
AF077028.1 | - | 2 | 186 | |||
AF115968.1 | - | 2 | 195 | |||
AK289579.1 | - | 2 | 180 | |||
AL080056.1 | - | 2 | 199 | |||
BC000466.2 | - | 2 | 180 | 2822781 | ||
BC019276.1 | - | 2 | 180 | 2822781 | ||
BM008944.1 | - | 2 | 136 | |||
BM686677.1 | - | 2 | 187 | |||
CF127298.1 | - | 2 | 178 | |||
CR533490.1 | - | 2 | 167 | |||
DA428446.1 | - | 2 | 239 |