Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204880.1 | NP_001191809.1 | 3 | 374 | ||
NM_003338.4 | NP_003329.1 | 4 | 406 | |||
XM_017016609.1 | XP_016872098.1 | 4 | 305 | |||
GenBank mRNA | AF257220.1 | - | 4 | 329 | ||
AJ272367.1 | - | 4 | 329 | |||
AK291901.1 | - | 4 | 377 | |||
BC005980.1 | - | 4 | 377 | 4106710 | ||
BC015997.1 | - | 4 | 381 | 4305900 | ||
BI598342.1 | - | 4 | 410 | |||
BT007041.1 | - | 4 | 183 | |||
DA618122.1 | - | 3 | 347 | |||
DA677507.1 | - | 4 | 406 | |||
X78140.1 | - | 4 | 198 |