Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003185.3 | NP_003176.2 | 6 | 1927 | ||
GenBank mRNA | AK123937.1 | - | 2 | 776 | ||
U75308.1 | - | 8 | 1513 | |||
Y11354.1 | - | 6 | 1921 |