Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284367.1 | NP_001271296.1 | 2 | 261 | ||
NM_001284368.1 | NP_001271297.1 | 2 | 170 | |||
NM_005004.3 | NP_004995.1 | 2 | 261 | |||
GenBank mRNA | AF044958.1 | - | 2 | 211 | ||
AF077028.1 | - | 2 | 208 | |||
AF115968.1 | - | 2 | 217 | |||
AK289579.1 | - | 2 | 202 | |||
AL080056.1 | - | 2 | 221 | |||
BC000466.2 | - | 2 | 202 | 2822781 | ||
BC019276.1 | - | 2 | 202 | 2822781 | ||
BM008944.1 | - | 2 | 158 | |||
BM686677.1 | - | 2 | 209 | |||
CF127298.1 | - | 2 | 200 | |||
CR533490.1 | - | 2 | 189 | |||
DA428446.1 | - | 2 | 261 |