Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098535.1 | NP_001092005.1 | 2 | 1243 | ||
NM_006604.2 | NP_006595.1 | 2 | 969 | |||
GenBank mRNA | AJ010232.1 | - | 2 | 1243 | ||
AK097738.1 | - | 3 | 1721 | |||
BC031689.1 | - | 2 | 969 | 5167710 | ||
CR456564.1 | - | 2 | 1085 |