Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006198.2 | NP_006189.2 | 3 | 230 | ||
GenBank mRNA | AF416716.1 | - | 3 | 178 | ||
AK289964.1 | - | 3 | 213 | |||
AM183336.1 | - | 3 | 168 | |||
BC013791.1 | - | 3 | 206 | 4345740 | ||
CR450319.1 | - | 2 | 149 | |||
U52969.1 | - | 3 | 230 | |||
X93349.1 | - | 3 | 207 |