Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012088.2 | NP_036220.1 | 2 | 381 | ||
XM_011527897.2 | XP_011526199.1 | 2 | 981 | |||
GenBank mRNA | AF091091.1 | - | 1 | 481 | ||
AJ243972.1 | - | 2 | 348 | |||
BC014006.2 | - | 2 | 355 | 4053022 | ||
BG764228.1 | - | 2 | 382 | |||
GQ472229.1 | - | 2 | 355 |