Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033925.1 | NP_001029097.1 | 3, 4 | 851 | ||
NM_001323964.1 | NP_001310893.1 | 3, 4 | 800 | |||
NM_001323965.1 | NP_001310894.1 | 3, 4 | 851 | |||
NM_001323967.1 | NP_001310896.1 | 3, 4 | 800 | |||
NM_001323968.1 | NP_001310897.1 | 4, 5 | 913 | |||
NM_001323969.1 | NP_001310898.1 | 4, 5 | 862 | |||
NM_001323970.1 | NP_001310899.1 | 4, 5 | 913 | |||
NM_003252.3 | NP_003243.1 | 3, 4 | 800 | |||
NR_136661.1 | 3, 4 | 800 | ||||
NR_136662.1 | 3, 4 | 851 | ||||
NR_136663.1 | 4, 5 | 862 | ||||
GenBank mRNA | AK290695.1 | - | 3, 4 | 753 | ||
AK290984.1 | - | 3, 4 | 450 | |||
AK291239.1 | - | 3, 4 | 433 | |||
AK295243.1 | - | 3, 4 | 245 | |||
AY633610.1 | - | 3, 4 | 244 | |||
BC030025.1 | - | 3, 4 | 805 | 4821759 | ||
D64015.1 | - | 3, 4 | 276 | |||
M96954.1 | - | 3, 4 | 281 |