Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145264.1 | NP_001138736.1 | 3 | 485 | ||
NM_014017.3 | NP_054736.1 | 4 | 575 | |||
GenBank mRNA | AF070659.1 | - | 4 | 487 | ||
AK307086.1 | - | 3 | 411 | |||
BC024190.2 | - | 4 | 451 | 2819969 | ||
BU184679.1 | - | 3 | 397 |