Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277197.1 | NP_001264126.1 | 3 | 273 | ||
NM_001277198.1 | NP_001264127.1 | 3 | 273 | |||
NM_001277199.1 | NP_001264128.1 | 3 | 551 | |||
NM_001277200.1 | NP_001264129.1 | 3 | 273 | |||
NM_014184.3 | NP_054903.1 | 3 | 273 | |||
NR_102347.1 | 2 | 254 | ||||
GenBank mRNA | AF161512.1 | - | 3 | 192 | ||
AK024569.1 | - | 188 | ||||
AK222634.1 | - | 3 | 178 | |||
AK289973.1 | - | 3 | 207 | |||
AK312064.1 | - | 3 | 198 | |||
AL136930.1 | - | 3 | 213 | |||
BC000573.1 | - | 3 | 188 | 3163724 | ||
BI752106.1 | - | 3 | 189 | |||
BQ051754.1 | - | 3 | 462 | |||
BX649076.1 | - | 3 | 177 | |||
DA185135.1 | - | 3 | 274 | |||
HY018269.1 | - | 2 | 170 | |||
HY145928.1 | - | 3 | 198 |