Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286102.1 | NP_001273031.1 | 2 | 235 | ||
NM_003269.4 | NP_003260.1 | 2 | 828 | |||
GenBank mRNA | AB307709.1 | - | 2 | 120 | ||
AF220532.1 | - | 2 | 827 | |||
AF411525.1 | - | 2 | 361 | |||
AK131541.1 | - | 2 | 235 | |||
BC028031.1 | - | 2 | 325 | 5242079 | ||
HQ692846.1 | - | 2 | 120 | |||
Y13276.1 | - | 2 | 288 |