Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020243.4 | NP_064628.1 | 2 | 231 | ||
GenBank mRNA | AB040119.1 | - | 2 | 213 | ||
AB041906.1 | - | 2 | 214 | |||
AF163256.1 | - | 2 | 214 | |||
AK024731.1 | - | 2 | 224 | |||
AK223021.1 | - | 2 | 229 | |||
AK312076.1 | - | 2 | 205 | |||
BC009363.2 | - | 2 | 206 | 4302531 | ||
CB142641.1 | - | 2 | 231 | |||
CR456475.1 | - | 2 | 206 |