Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021026.2 | NP_066306.2 | 1 | 478 | ||
XM_011530298.2 | XP_011528600.1 | 3 | 438 | |||
XM_011530299.2 | XP_011528601.1 | 2 | 340 | |||
XM_017028900.1 | XP_016884389.1 | 5 | 825 | |||
XM_017028901.1 | XP_016884390.1 | 4 | 759 | |||
XM_017028902.1 | XP_016884391.1 | 9 | 3803 | |||
GenBank mRNA | AJ010228.1 | - | 1 | 478 | ||
AJ010229.1 | - | 1 | 478 | |||
BC104768.1 | - | 1 | 208 | |||
CR456562.2 | - | 1 | 300 |