Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020309.3 | NP_064705.1 | 3, 4 | 607 | ||
GenBank mRNA | AB032436.1 | - | 3, 4 | 456 | ||
AK057879.1 | - | 553 | ||||
AK095659.2 | - | 2, 3 | 1886 | |||
AK289622.1 | - | 15 | ||||
AK294226.1 | - | 3, 4 | 413 | |||
AK294405.1 | - | 3, 4 | 456 | |||
BC059379.1 | - | 3, 4 | 582 | 5262415 | ||
BG701511.1 | - | 3, 4 | 583 |