Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRABD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320484.1 | NP_001307413.1 | 8 | 815 | ||
NM_001320485.1 | NP_001307414.1 | 8 | 803 | |||
NM_001320487.1 | NP_001307416.1 | 8 | 837 | |||
NM_001320488.1 | NP_001307417.1 | 8 | 837 | |||
NM_025204.3 | NP_079480.2 | 8 | 828 | |||
NR_135275.1 | 8 | 799 | ||||
XM_006724422.3 | XP_006724485.1 | 8 | 807 | |||
XM_006724423.3 | XP_006724486.1 | 8 | 800 | |||
XM_011530712.2 | XP_011529014.2 | 8 | 875 | |||
XM_011530713.2 | XP_011529015.2 | 8 | 873 | |||
XM_011530715.2 | XP_011529017.2 | 8 | 878 | |||
XM_011530716.2 | XP_011529018.1 | 8 | 838 | |||
XM_011530717.2 | XP_011529019.1 | 8 | 819 | |||
XM_011530718.2 | XP_011529020.1 | 8 | 798 | |||
XM_017028943.1 | XP_016884432.1 | 8 | 880 | |||
XM_017028944.1 | XP_016884433.1 | 8 | 915 | |||
GenBank mRNA | AF218001.1 | - | 1 | 963 | ||
AK024447.1 | - | 1 | 3747 | |||
AK292050.1 | - | 8 | 807 | |||
AL449244.1 | - | 8 | 809 | |||
AY203945.1 | - | 8 | 795 | |||
BC012445.1 | - | 8 | 647 | 3857552 | ||
BC093029.1 | - | 8 | 822 | 30527560 | ||
BX389277.2 | - | 2 | 95 |