Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF45 SIRNA Assays ›
Gene Symbol:
ZNF45
Gene Name:
zinc finger protein 45
Gene Aliases:
KOX5, ZNF13
Chromosome Location:
Chr.19: 43912624 - 43935278 on Build GRCh38
UniGene ID:
Hs.381285
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4554
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF45
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003425.3
NP_003416.1
9, 10
1334
XM_011527267.1
XP_011525569.1
10, 11
1417
XM_011527269.1
XP_011525571.1
9, 10
1330
XM_011527271.1
XP_011525573.1
9, 10
1047
XM_011527273.1
XP_011525575.1
8, 9
1247
XM_017027217.1
XP_016882706.1
11, 12
1536
XM_017027218.1
XP_016882707.1
11, 12
1532
XM_017027219.1
XP_016882708.1
10, 11
1414
XM_017027220.1
XP_016882709.1
11, 12
1248
XM_017027221.1
XP_016882710.1
11, 12
1550
XM_017027222.1
XP_016882711.1
10, 11
1453
XM_017027223.1
XP_016882712.1
11, 12
1277
XM_017027224.1
XP_016882713.1
9, 10
1352
XM_017027225.1
XP_016882714.1
10, 11
1165
XM_017027226.1
XP_016882715.1
10, 11
1161
XM_017027227.1
XP_016882716.1
8, 9
959
XM_017027228.1
XP_016882717.1
9, 10
1397
XM_017027229.1
XP_016882718.1
10, 11
1516
XM_017027230.1
XP_016882719.1
9, 10
1393
GenBank mRNA
AK127739.1
-
9, 10
1334
BC037575.1
-
6, 7
601
5539981
L75847.1
-
3, 4
346
M67509.1
-
3, 4
346
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
multicellular organism development
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0