Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168399.1 | NP_001161871.1 | 3 | 1000 | ||
NM_001168400.1 | NP_001161872.1 | 3 | 997 | |||
NM_001168401.1 | NP_001161873.1 | 3 | 742 | |||
NM_032621.3 | NP_116010.1 | 3 | 814 | |||
GenBank mRNA | AF251053.1 | - | 3 | 646 | ||
AY833560.1 | - | 3 | 746 | |||
BC015522.1 | - | 3 | 692 | 3914401 |