Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFIKKN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053284.2 | NP_444514.1 | 2 | 442 | ||
GenBank mRNA | AF422194.1 | - | 2 | 200 | ||
AK075356.1 | - | 2 | 522 | |||
AK128777.1 | - | 1 | 3370 | |||
AK314757.1 | - | 2 | 504 | |||
BC101602.1 | - | 2 | 283 | 8069108 | ||
BC101606.1 | - | 2 | 283 | 8069112 | ||
CN425794.1 | - | 2 | 453 |