Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000863.2 | NP_000854.1 | 1 | 694 | ||
GenBank mRNA | AK290080.1 | - | 1 | 445 | ||
AK315325.1 | - | 1 | 445 | |||
BC069065.1 | - | 1 | 75 | 7216878 | ||
BC096206.1 | - | 1 | 75 | |||
BC096207.1 | - | 1 | 75 | 40004711 | ||
BC096208.1 | - | 1 | 75 | |||
KJ534870.1 | - | 1 | 75 | |||
M81590.1 | - | 1 | 136 |
Set Membership: |
G-protein coupled receptor |