Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002904.5 | NP_002895.3 | 6, 7 | 614 | ||
XM_011514913.2 | XP_011513215.1 | 6, 7 | 554 | |||
GenBank mRNA | AK300898.1 | - | 6, 7 | 489 | ||
AK302652.1 | - | 6, 7 | 601 | |||
BC025235.1 | - | 6, 7 | 465 | 4876045 | ||
BC050617.2 | - | 6, 7 | 481 | 6168937 | ||
JQ313904.1 | - | 5, 6 | 410 | |||
L03411.1 | - | 6, 7 | 496 | |||
X16105.1 | - | 3, 4 | 280 |