Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003185.3 | NP_003176.2 | 11 | 2734 | ||
GenBank mRNA | AK123937.1 | - | 7 | 1583 | ||
U75308.1 | - | 13 | 2320 | |||
Y11354.1 | - | 11 | 2728 |