Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308353.1 | NP_001295282.1 | 3 | 330 | ||
NM_005855.3 | NP_005846.1 | 2 | 269 | |||
XM_017003152.1 | XP_016858641.1 | 2 | 191 | |||
XM_017003153.1 | XP_016858642.1 | 2 | 499 | |||
XM_017003154.1 | XP_016858643.1 | 2 | 276 | |||
XM_017003155.1 | XP_016858644.1 | 2 | 197 | |||
XM_017003156.1 | XP_016858645.1 | 2 | 217 | |||
GenBank mRNA | AI017108.1 | - | 2 | 269 | ||
AJ001014.1 | - | 2 | 169 | |||
AY265457.1 | - | 2 | 137 | |||
BC000548.2 | - | 2 | 161 | 3163522 | ||
BI548986.1 | - | 3 | 256 | |||
CR542032.1 | - | 2 | 137 | |||
CR542044.1 | - | 2 | 137 |